Klinger Laser Ray Box, Electronic

Supplier: Klinger
LRB-ELEC-KL
470346-482EA 621.2 USD
470346-482
Klinger Laser Ray Box, Electronic
Educational Materials Physics Educational Materials Optics Learning Activities
This laser ray box has several modes, with 1, 3, or 5 beams.

  • Multiple ray configurations options: five rays (1, 2, 3, 4, 5) - three rays (1, 3, 5) - three rays (2, 3, 4) - single ray (3)
  • Powered by 2×AA batteries or included 3 V/300 mA power supply
  • Laser type: diode wavelength, 635 nm
  • Class II Laser, <1 mW

The electronic laser ray box consists of five independent red laser modules producing five parallel lines that are collimated by cylindrical lenses to be highly visible. This electronic version adds electronic shade control for each of the beams. It is taped with magnetized foil, allowing intuitive attachment to a magnetic board. Use the ON/OFF switch to select between one ray, three central rays, three alternating rays, or all five rays depending on specific needs.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR