HOBO® No-Lead Water Flow Meter Sensor
Supplier: ONSET COMPUTER CORPORATION SE
T-MINOL-130-NL
470346-448EA 402.05 USD
470346-448
HOBO® No-Lead Water Flow Meter Sensor
Electrodes Other Electrodes
A compact, affordable water meter to measure cold and hot water flow rates.
  • Measures cold and hot water flow rates
  • Suitable for use in apartments, assisted-living communities, and other types of commercial buildings
  • Configured for use with 3/4" pipes; use step-downs for smaller pipe
Designed for deployment in apartments, assisted living communities, and other commercial buildings.