HOBO® AC Current Switch Sensor
Supplier: ONSET COMPUTER CORPORATION SE
CSV-A8
470346-426EA 163.79 USD
470346-426
HOBO® AC Current Switch Sensor
Electrodes Other Electrodes
Measures equipment runtime (time-of-use).
  • Installs around a current-carrying wire to provide a relay signal indicating equipment on/off
  • Adjustable threshold provides maximum application flexibility
  • UL listed
Provides an adjustable current threshold of 0.5 to 175 A. Simply install the sensor around a current-carrying wire to the equipment to be monitored and then connect the sensor to a compatible data logger.
Ordering information: Compatible with HOBO UX90 and UX120-017 data loggers.