Thin-Walled PCR Tubes, 0.2 ml
Supplier: SUPERTEK SCIENTIFIC LLC
A10279/1
470346-260CS 128.19 USD
470346-260
Thin-Walled PCR Tubes, 0.2 ml
Tubes PCR Tubes
A thin-walled tube for efficient thermal transfer in PCR or other micro-reactions.
  • PCR tube made of prime virgin polypropylene
  • Transparent, soft and robust
  • High light transmittance for optimal performance