TraxIt® Single Use Thermometers
Supplier: MEDICAL INDICATORS,INC
1313-20
470346-244BX 74.39 USD
470346-244
TraxIt® Single Use Thermometers
Thermometers Digital Thermometers
The ideal thermometer for pediatric care.
  • Single patient use
  • 48 hour continuous read
TraxIt® thermometers are meant to simplify the process of taking a child’s temperature. Instead of having to repeatedly take a child’s temperature, the TraxIt® adheres to the child’s underarm and continuously reads the child’s temperature for up to 48 hours. TraxIt®’s adhesive is a gentle, latex-free, waterproof, hypoallergenic medical tape that will not irritate a child’s healthy skin.