Insect Spreading Board
Supplier: ARCO
4120300
470346-234EA 20.14 USD
470346-234
Insect Spreading Board
Specimen Mounting Supplies
Prepare your insects for mounting.
  • Easily adjustable width
  • Cork coating
Made of wood covered with thick cork sheet. The groove from 0 to 12 mm is smoothly adjustable, die-pressed adjustment chrome plated brass fittings.