UniStore™ Amber Reagent Bottles, Narrow Mouth, HDPE
Supplier: United Scientific Supplies
33421 33422 33423 33424 33425 33426 33427 33428 33429
470346-198EA 29.1 USD
470346-198 470346-182 470346-150 470346-126 470346-196 470346-190 470346-134 470346-130 470346-142
UniStore™ Amber Reagent Bottles, Narrow Mouth, HDPE
Bottles Plastic Bottles
Leakproof, amber laboratory quality bottles with many applications in the field or laboratory.
  • Protect light-sensitive materials during shipping and storage
  • Bottles meet USP requirements for storage of light-sensitive materials
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact
  • Free of heavy metals, with leakproof closure featuring an integrated seal ring
  • Precision molded for consistent wall thickness and smooth interior and exterior surfaces
UniStore™ narrow mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. The amber bottles are designed to reduce UV light transmission to protect light-sensitive contents, and conform to USP requirements for storage of light-sensitive materials. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.