Eisco Alcohol Lamp
Supplier: Wards
CH0533
470346-104EA 9.15 USD
470346-104
Eisco Alcohol Lamp
Heaters Spirit Lamps
Alcohol lamp with a polyhedral shape for positioning flame at different angles.
  • Traditional alcohol lamp with 120 ml glass tank
  • Features nickel-plated cap and fiber wick
  • Polyhedral shape allows for positioning flame at multiple angles
  • Use with 95% denatured alcohol as fuel
Glass alcohol lamp with metal cap and fiber wick has a 120 ml fuel capacity. Safer alternative to Bunsen and propane burners for basic laboratory applications. Sufficient for performing standard biology and sterilization procedures. Use with 95% denatured ethyl alcohol. Trim wick to no more than 2 cm.