Porcelain Combustion Boat with Handle
Supplier: Wards
CH0751
470346-002EA 7.75 USD
470346-002
Porcelain Combustion Boat with Handle
Boats Combustion Boats
Measuring 4×1" exterior and 3×¹/₂" interior, these porcelain boats with handles come in a pack of 10 and provide a reliable container for combustion experiments.
  • Pack of 10 porcelain combustion boats with handle
  • Made from porcelain for increased resistance to heat and caustic chemicals
  • Interior measures approximately 3×1"
  • Each pack comes with 10 individual porcelain combustion boats
  • Great for any classroom or laboratory