Eisco Bunsen Burner with Flame Stabilizer
Supplier: Wards
CH0093A CH0093B
470343-848EA 28.45 USD
470343-848 470345-988
Eisco Bunsen Burner with Flame Stabilizer
Heaters Gas Burners
Standard-feature option for variety of lab applications.
  • High-quality Bunsen burner, suitable for a variety of laboratory applications
  • Aluminum burner mixing tube with brass flame stabilizer
  • Serrated inlet arm accepts 6 to 8 mm tubing
The 13 mm Ø aluminum burner mixing tube comes complete with a fixed brass flame stabilizer and is threaded into a die casted zinc alloy base. Air supply is controlled by a nickel plated brass sleeve covering ports at the lower end of the mixing tube.
Overall size: Burner 6.3 tall × 3" base Ø