Eisco Drechsel Gas Washing Bottles
Supplier: Wards
CH0196B CH0196A CH0196C
470344-412EA 43.3 USD
470344-412 470345-952 470345-980
Eisco Drechsel Gas Washing Bottles
Bottles Gas Washing Bottles
Remove impurities from gases.
  • Made from high quality borosilicate 3.3 glass
  • Interchangeable ground glass joint allows for easy sealing and exchange
  • Both bottle and joint head are autoclavable
Autoclavable, with superior chemical durability. Low temperature gradient; can withstand high temperatures and thermal shocks.