Gooch Crucibles
Supplier: Wards
CH0509EPK5
470345-938EA 20.1 USD
470345-938
Gooch Crucibles
Crucibles
Set of 5 glazed porcelain Gooch crucibles with a 30 ml capacity, featuring an unglazed rim and bottom surface, ensuring accurate filtration and sample preparation for various laboratory applications.
  • Crucibles are made of porcelain and have a perforated bottom
  • Each crucible is glazed inside and outside with the exception of outside bottom surface and rim
  • Top outside diameter 42 mm, overall height 40 mm, 2 mm thickness
  • 30 ml capacity
Pack of 5, 30 ml capacity porcelain filtering crucibles.