Eisco Small Beaker Tongs
Supplier: Wards
BKTGMINI
470345-918EA 5.6 USD
470345-918
Eisco Small Beaker Tongs
Tongs
Ideal for gentle handling of smaller sized glassware.
  • Handle made of high-quality, nickel-plated steel
  • Curved jaws with rubber coated tips for added grip strength and safety
  • For use with 25 to 100 ml capacity beakers
Small metal beaker tongs made of nickel-plated steel. Tongs measure 7.25" in length and are 2.5" wide.