Eisco® Specific Gravity Bottle
Supplier: Wards
PH0122B
470345-862EA
13
USD
470345-862
Eisco® Specific Gravity Bottle
Specific Gravity Bottles
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...