H-Shaped Retort Base
Supplier: Wards
CH0656
470345-860EA 74.45 USD
470345-860
H-Shaped Retort Base
Support Stands
H-shaped retort base with an 8×8" heavy-duty cast iron base in blue. The base ensures stability and strength for retort stands and setups in laboratory environments.
  • H-shaped frame retort base made of chemical-resistant cast iron with a blue coating
  • The four-point base weighs 7.3 lbs. providing superb stability for experiments and setups
  • Base accommodates 11 mm (0.4") rods (sold separately) secured with a tightening screw
  • Durable and sturdy construction of this set makes it an excellent choice for science classrooms
H-frame retort base lab stands areMore Product Information