Eisco® Large Vernier Caliper Demonstration, 24"
Supplier: Wards
PH0077
470345-656EA 33 USD
470345-656
Eisco® Large Vernier Caliper Demonstration, 24"
Calipers
Large 24" long, dual scale, metric and imperial graduations, designed for demonstrations, precise and accurate measurements in workshops and classrooms.
  • Large Vernier caliper demonstration made from quality wood
  • Each Vernier demonstration is 24" long, 7" wide, 0.75" thick, and weighs 2.4 pounds
  • The sliding arm is approximately 7" long and slides smoothly up and down the main arm
This Large Vernier Caliper Demonstration is an excellent visual aid for teaching students how to accurately take and read measurements using a Vernier caliper.