12 V DC Power Supply
Supplier: Wards
CH0740E
470345-638EA 161.85 USD
470345-638
12 V DC Power Supply
Power Supplies
With CE certification, this power supply is specifically designed for use with Hoffman voltmeters, ensuring accurate and reliable readings in electrolysis experiments.
  • Power supply for Hoffmann voltmeter
  • 12 V DC 2 amp main input 120 V, output wattage 24 W max. input wattage 28 W max. Fuse: F0.5A/250V
  • Heavy duty gray case
  • Includes 120 V power cord