Eisco Circular Tripod Stands, Cast Iron
Supplier: Wards
CH0644A CH0644B
470345-610EA 14.55 USD
470345-610 470345-726
Eisco Circular Tripod Stands, Cast Iron
Tripods
Circular tripod stands with zinc-plated cast iron tops for supporting objects over Bunsen burners.
  • Tripod stand for suspending objects to be heated above a Bunsen burner
  • Durable zinc-plated cast iron ring
  • Legs made of mild steel with rubber traction caps to prevent slippage
  • Legs are detachable for convenient storage
Rugged circular tripod stands have a zinc-plated iron ring and sturdy steel legs that are detachable for storage. Legs come with rubber traction caps on them.