Eisco® Slotted Mass Set with Hanger, Stainless Steel, 9 Weights, 500 g
Supplier: Wards
PH0258NSS
470345-566EA 47.45 USD
470345-566
Eisco® Slotted Mass Set with Hanger, Stainless Steel, 9 Weights, 500 g
Weights Calibration Weights
Set includes nine weights totaling 500 g, made of stainless steel for durability. Set inclues a hanger for suspending.
  • Stainless steel hanger with hook end for suspending masses in force experiments
  • Masses can be used for studying pressure, strength, or torque in the classroom or lab
  • Masses can be used for calibrating scales and balances
Eisco Labs stainless steel slotted masses set can be used with hanging balances as well as for calibration and studying pressure, strength, or torque in the classroom or lab. Unit includes nine weights, and one hanger, each weighing 50 g. Total weight of set is 500 g.