Heavy-Duty Cork Boring Machine
Supplier: Wards
CH0326A
470345-462EA 423 USD
470345-462
Heavy-Duty Cork Boring Machine
Cork Borers
Sturdy and robust cork boring machine designed for heavy-duty usage, ensuring efficient and precise cork-boring tasks.
  • 12 nickel-plated borers included
  • Cutting sizes 7 to 25 mm
  • Hand crank
  • 4 pre-drilled holes in base for mounting to a table top
The cork boring machine is designed for drilling uniform and straight holes in cork or rubber stoppers. Cast iron frame with wood platform (10 cm diameter) for support. Adjustable height. Completed with set of 12 nickel-plated borers. Base plate has 4 mounting holes to keep boring machine stable during use.