Plastic Single Burette Clamp
Supplier: Wards
CH0262A
470345-314EA 4.2 USD
470345-314
Plastic Single Burette Clamp
Clamps General Purpose Clamps
Single polypropylene burette clamp.
  • Single clamp - fits rods 10 to 15 mm in diameter
  • Features two tapered wedges and a spring-loaded jack
  • Made of high quality polypropylene
The sturdy, polypropylene single burette clamp is a great tool for tightly holding 10 and 12.5 mm rods. The spring-loaded jack and two tapered wedges assure a snug and safe fit.