Laboratory Hydrometer

Supplier: Wards
PH0132B
470345-280EA 12.45 USD
470345-280
Laboratory Hydrometer
Hydrometers
Specific gravity measurement for heavy liquids, range of 1.500 to 2.000×0.010, comes with a protective case.

  • Dual scale hydrometer for measuring the relative density in heavy liquids
  • Shot loaded
  • Designed for laboratory use, but can be used in homes and garages

Delivery information: Includes sturdy, plastic protective case.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR