Eisco Natural Fiber Cylinder Brush with Fan-Shaped End
Supplier: Wards
CH0220A
470345-238EA 10.9 USD
470345-238
Eisco Natural Fiber Cylinder Brush with Fan-Shaped End
Brushes
Made for cleaning graduated cylinders, bottle necks, and more.
  • Medium-soft fiber bristles
  • Stainless steel, twisted wire handle that is resistant to corrosion
  • Fanned end for reaching every surface and corner
  • Moderate flexibility allows easy modification to handle shape
This medium soft fiber bristle brush is perfect for cleaning hard to reach, narrow spaces such as graduated cylinders, bottle necks, and more. The fanned end allows you to easily clean rounded surfaces and reach all corners. The stainless steel, twisted handle is semi-flexible, allowing you to modify the shape as needed. Looped end for easy hanging. Brush measures 12.5" in total length, with the bristle section measuring 5" in length and 1.5" in diameter, and handle measuring 7.5". Fits inside items with diameters of 1.25 to 1.5".