Stainless Steel Spirit Lamp
Supplier: Wards
CH0535B
470345-154EA 15.6 USD
470345-154
Stainless Steel Spirit Lamp
Heaters Spirit Lamps
Stainless steel spirit lamp, provides a reliable and safe heat source for various laboratory applications.
  • 80 ml capacity
  • Safely produces open flame for basic laboratory activities
  • Wick controlled by simple ratched knob for ease of use
Stainless steel spirit lamp is an essential piece of laboratory equipment used to safely produce an open flame with a shorter height and lower temperature than that of a Bunsen burner. The flame is typically hot enough to sterilize other equipment and is adequate for typical molecular biology activities.
Delivery information: Supplied with chain-attached extinguisher cap.