Eisco Clay Pipe Triangles
Supplier: Wards
CH0530B CH0530A CH0530C
470344-166EA 1.85 USD
470344-166 470344-366 470345-128
Eisco Clay Pipe Triangles
Tripods
Braided iron wire wrapped with insulating clay piping on three sides, for supporting hot objects.
  • Holds objects like crucibles on ring stands during or after high heating
  • Made of braided iron wire wrapped with clay piping on three sides
  • Piping measures 11 mm in diameter
Clay Pipe Triangles of varying sizes for supporting very hot items. Made of braided iron wire wrapped with clay piping on its three sizes.