Heavy-Duty Beaker Tongs
Supplier: Wards
CH0731
470345-086EA 15.75 USD
470345-086
Heavy-Duty Beaker Tongs
Tongs
These 12" long stainless steel tongs with cork-lined jaws and a riveted joint are ideal for handling hot beakers ranging from 100 to 1500 ml capacities.
  • Heavy-duty beaker tongs made from stainless steel
  • Designed with a riveted joint for increased strength
  • Cork-lined jaws for use with hot beakers and glassware
  • Measures approximately 12" long, 4.25" wide
Heavy-duty beaker tongs feature a riveted joint for increased strength, and a cork-lined jaw for handling hot beakers and glassware. Permits handling of hot beakers from 100 to 1500 ml capacities.