3 Prong Rubber Coated Clamp
Supplier: Wards
CH0684B
470344-984EA 21.3 USD
470344-984
3 Prong Rubber Coated Clamp
Clamps General Purpose Clamps
Rubber-coated 3-prong clamp with chemical-resistant prongs, featuring a maximum opening of 4.5" (11.4 cm) and a total length of 10.5" (26.7 cm), providing a secure and stable grip for a wide range of objects and applications.
  • Overall length of 10.5" (26.7 cm)
  • Maximum clamp opening of 4.5" (11.4 cm)
  • Features rubber coated, chemical resistant prongs
  • All metal construction with threated steel actuating joints
  • Attached to steel connecting rod. Steel rod measures 6" long, 0.25" diameter.
Three prong universal clamp with rubber coated, chemical resistant prongs. Attached to a steel 0.25" diameter rod. All metal construction with threated steel actuating joints.