Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 500 g Total
Supplier: Wards
PHZNW500SET
470344-944EA 30.45 USD
470344-944
Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 500 g Total
Weights Calibration Weights
Set of 9 zinc-casted slotted masses with one hanger each of 50 g, total weight of 500 g. For use in physics experiments and demonstrations.
  • Powder-coated, die casted zinc
  • 50 g slotted weights with accuracy ±2%
  • Complete with hanger
  • Great alternative to expensive brass sets
Beautiful die casted zinc 9-piece slotted masses set on durable hanger. Each weight is 50 g, powder-coated, with a ±2% accuracy.