Universal Laboratory Hydrometer
Supplier: Wards
PH0128
470344-922EA 10.4 USD
470344-922
Universal Laboratory Hydrometer
Hydrometers
Measures specific gravity range of 0.700 to 2.000 with 0.01 graduations and dual scale for light and heavy liquids.
  • Universal laboratory hydrometer in hard plastic case measure specific gravity range 0.700 to 2.000
  • Dual scale hydrometer designed for lab use
  • Can be used for beer, wine, kombucha, mead, and other specific gravity activities
  • Designed to be used in the laboratory, but can be used in the home or garage as well
Lab quality hydrometer with dual scale.