Lattice Support Clamps
Supplier: Wards
CH0664C
470344-874EA 50.35 USD
470344-874
Lattice Support Clamps
Clamps General Purpose Clamps
Lattice support clamps, sold in a pack of 12, designed to fit rods up to 12.5 mm in diameter, providing stability and versatility in securing apparatus for experiments and demonstrations.
  • Pack of 12 lattice support clamps that fit rods up to 12.5 mm in diameter
  • Clamps are made of high quality metal with a silver finish
  • Tighten clamps using an Allen wrench - Avoid overtightening
  • Twelve clamps only - rods pictured are not included
  • Clamps measure 29×28 mm with an inner diameter of 13 mm