Eisco Bunsen Burner with Needle Valve, Natural Gas
Supplier: Wards
CH0091B
470344-784EA 47.5 USD
470344-784
Eisco Bunsen Burner with Needle Valve, Natural Gas
Heaters Gas Burners
Easy flame adjustment.
  • Needle valve gas flow control knob fitted above the base
  • Suitable for use with natural gas (methane)
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Circular, pressure die casted base
Durable and efficient, excellent for general laboratory applications. Suitable for use with natural gas. Burner comes equipped with a serrated inlet arm that measures 8 mm in Ø and 1" in length. Gas flow is controlled by needle valve fitted above the base, permitting easy flame adjustment with minimizing possibility of tipping. Burner measures 5.75" tall, and the base measures 3" in Ø.