Advanced Organic Chemistry Distillation Glassware Set
Supplier: Wards
CH0878
470344-758EA 273.29 USD
470344-758
Advanced Organic Chemistry Distillation Glassware Set
Distillation Components Distillation Sets
This 17-piece distillation glassware set offers 22 interchangeable fittings, making it ideal for advanced organic chemistry experiments, all crafted from high-quality borosilicate glass for superior performance and durability.
  • The perfect set for organic chemistry labs, distillation, fractional distillation, separation, purification, and synthesis
  • All pieces made of 3.3 borosilicate glass that is hand blown and polished for perfect fit
  • Eisco E-Lab glass guarantee, any piece breaks during shipping will be replaced free of charge
  • Metal ware stands not included in set
Set consists of: Liebig condenser, dropping funnel, deduction adapter, air leak/steam inlet, receiver adapter, multiple adapter, 250 ml Erlenmeyer flask, 250 ml round bottom flask, 100 ml round bottom flask, 50 ml round bottom flask, 2 Stoppers (19/26), socket cone, 1 stopper (24/29), thermometer. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.