Platinum Wire
Supplier: Wards
CH0538
470344-750EA 13.09 USD
470344-750
Platinum Wire
Wire
Platinum wire supplied in a glass tube, designed specifically for flame tests, ensuring accurate and consistent results.
  • Platinum wire ideal for flame test
  • Supplied in borosilicate glass tube, fused into glass handle
  • Attached to silicon cork
  • Tube measures 5" long and 0.6" in diameter. Wire measures 18 to 20 mm long and approximately 0.12 mm in diameter