Laboratory Jacks, Steel
Supplier: Wards
CH0642A CH0642B
470344-714EA 115.6 USD
470344-714 470345-660
Laboratory Jacks, Steel
Laboratory Jacks
Height-adjustable lightweight lab jacks offer strong, stable support for laboratory apparatus.
  • Adjustable and reversible Lab Jacks with screw-operated mechanism
  • Made of powder coated lightweight steel resistant to many chemicals
  • Single piece base provides enhanced torsional rigidity for stabilization under load
  • Can be autoclaved and chemically cleaned
Strong, stable support for laboratory apparatus. The lab jack has a screw-operated scissor action mechanism for easy, fluid movement of platform - with and without load - and precise height adjustments. For physical, chemical, biological experiments and other uses.