Eisco LDPE Dropping Bottles, Pack of 12
Supplier: Wards
CH0182CPK12 CH0182BPK12 CH0182EPK12 CH0182APK12 CH0182DPK12
470343-740EA 44.9 USD
470343-740 470344-682 470345-586 470346-044 470346-088
Eisco LDPE Dropping Bottles, Pack of 12
Bottles Dropping and Dispensing Bottles
Dispenses precise uniform drops of liquid.
  • Made of translucent low density polyethylene plastic (LDPE)
  • Screw cap with a built in dropper nozzle allows for small, uniform drops of liquid
  • Attached cover for nozzle prevents spills
Nozzle cover teathered to cap prevents leakage and spills.
Packaging: Comes in packs of 12 bottles.