Eisco® 9-Piece Retractable Metric Hooked Weight Set, Stainless Steel
Supplier: Wards
PH0037DSSECO
470344-680EA 96.3 USD
470344-680
Eisco® 9-Piece Retractable Metric Hooked Weight Set, Stainless Steel
Weights Calibration Weights
Set of 9 stainless steel hooked metric weights with retractable hooks, packed in a premium wooden storage block.
  • Includes 9 individually calibrated hooked weights totaling 2100 g: (1) 10 g, (2) 20 g, (1) 50 g, (1) 100 g, (2) 200 g, (1) 500 g and (1) 1000 g
  • Made from high-grade stainless steel
  • Retractable hooks on top and bottom of each piece allow for chaining together or sitting flat on a surface
  • Weights are printed on each piece for easy identification
  • Includes polished wooden storage block with a place for each hooked weight
This 9-piece hooked weights set is made from high quality stainless steel and includes a wooden storage block. Hooked weights each have retractable hooks, on the top and bottom of the main body (with the exception of the 10 g weight, which has only a hook on the top of the main body), allowing pieces to be hooked in tandem, as well as allowing each piece to sit flat on a surface. Includes weights ranging 10 to 1000 metric grams. Hooked weights include (1) 10 g, (2) 20 g (1) 50 g, (1) 100 g, (2) 200 g, (1) 500 g, (1) 1000 g. Wooden block measures 8" wide, 3.75" long and 3" tall.