Burette/Tube Clamp
Supplier: Wards
CH0258A
470344-546EA 19.39 USD
470344-546
Burette/Tube Clamp
Clamps General Purpose Clamps
Single burette/tube clamp made of high-strength alloy, equipped with spring-loaded jaws, has a width capacity of ⁷/₈", holds burettes up to 100 ml, and fits maximum rod diameter of 16 mm.
  • Spring loaded, high strength alloy jaws holds objects in place
  • Powder coated for long lasting finish
This burette clamp made from high strength alloy has spring-loaded jaws suitable for all burettes up to 100 ml. Can be mounted on rods up to 16 mm diameter.