Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 100 g Total
Supplier: Wards
PHZNW100SET
470344-466EA 12.25 USD
470344-466
Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 100 g Total
Weights Calibration Weights
Set of 9 zinc-casted slotted masses with one hanger each of 10 g, total weight of 100 g. For use in physics experiments and demonstrations.
  • Powder-coated, die casted zinc
  • 10 g slotted weights with accuracy ±2%
  • Complete with hanger
  • Great alternative to expensive brass sets
Beautiful die casted zinc 9-piece slotted masses set on durable hanger. Each weight is 10 g, powder-coated, with a ±2% accuracy.