Eisco Glass Crystallizing Dishes
Supplier: Wards
CH0066I CH0066EPK6
470344-102EA 35.2 USD
470344-102 470344-370
Eisco Glass Crystallizing Dishes
Trays/Dishes/Pots Crystallizing Dishes
Designed for crystallization and storage.
  • Crystallizing dish made of high quality, heavy duty borosilicate 3.3 glass
  • Features a flat bottom, beaded rim, and a spout for easy pouring
  • Superior chemical durability and heat tolerance
  • Can be safely sterilized in autoclaves at 121 °C
100 ml capacity crystallizing dishes made of high quality, heavy duty borosilicate 3.3 glass. Features flat base, beaded rim, and spout for easy pouring. Autoclavable, with superior chemical durability. Low temperature gradient can withstand high temperatures and thermal shocks.