Boss Head Clamp, Powder Coated
Supplier: Wards
CH0663AP
470344-312EA 7.69 USD
470344-312
Boss Head Clamp, Powder Coated
Clamps General Purpose Clamps
Boss head clamp made of die-cast alloy with powder-coated finish, suitable for rods up to 16 mm, offering sturdy and reliable support for lab equipment.
  • Aluminum alloy clamp
  • Metal clamping screws with plastic heads
  • Accommodates round rods up to 16 mm in diameter
For use with laboratory stands, this popular boss head is made of die casted alloy. It accepts rods up to 16 mm in diameter at a right angle to one another. Metal thumbscrews with blue plastic heads allow for adjustment. Measures approximately 11.7×3×3 cm in size.