Heavy-Duty Flask Tongs
Supplier: Wards
CH0733
470344-264EA 14.3 USD
470344-264
Heavy-Duty Flask Tongs
Tongs
Made of stainless steel and measuring 10" long, these tongs with cork-lined jaws offer a strong grip for secure handling of flasks in laboratory settings.
  • Heavy-duty flask tongs made from stainless steel
  • Square opening for miscellaneous applications
  • Cork-lined jaws to use with hot flasks and glassware
  • Permits handling of hot flasks and objects
Heavy-duty flask tongs feature a cork-lined jaw for handling hot flasks and glassware and have a square opening for miscellaneous applications. Permits handling of hot flasks and glassware. The flask tongs measure approximately 10" long and 4.25" wide when closed. Reliable heavy-duty build and design.