Eisco Plastic Spot Plate, 12-Well
Supplier: Wards
CH0542C
470344-212EA 5.3 USD
470344-212
Eisco Plastic Spot Plate, 12-Well
Staining Tiles
Spot plate made of high quality polyethylene plastic which can be cleaned and reused.
  • Spot plate made of high quality polyethylene, with excellent chemical resistance
  • Features 12 wells measuring 22 mm in diameter
  • Can be cleaned and reused
  • Perfect for using with liquid or small solid samples
It can hold approximately 1.5 ml. Useful for comparing results among students in a lab, or in classroom demonstrations.
Height: 0.4"