Eisco Test Sieves/Strainers, Set of 6, ASTM
Supplier: Wards
BI0139
470344-206EA 145.8 USD
470344-206
Eisco Test Sieves/Strainers, Set of 6, ASTM
Sieves Test Sieves
Plastic frames with stainless steel mesh for serial sifting of samples.
  • Six plastic sieves with stainless steel mesh, cover and bottom collector
  • Complete with plastic lid and bottom collector
  • Stackable
Each sieve has a plastic frame with a single mesh size (ASTM mesh no. 5, 10, 35, 60, 120, 230). Total height of stacked sieves: 15.25". Not foodsafe.