Eisco Plastic Stirring Rods, Pack of 12
Supplier: Wards
CH0757E
470344-172EA 16.4 USD
470344-172
Eisco Plastic Stirring Rods, Pack of 12
Stirrers Stirring Rods
Stirring rods with rounded ends, made of chemically-resistant polypropylene plastic.
  • Pack of 12 multipurpose stirring rods with rounded ends
  • Made of high quality, heavy-duty polypropylene
  • Autoclavable with very good resistant to chemicals
  • Each rod measures 12" in length, and is 0.25" in Ø