Eisco Bunsen Burner with Flame Stabilizer
Supplier: Wards
CH0087B CH0087A
470343-554EA 30.35 USD
470343-554 470344-146
Eisco Bunsen Burner with Flame Stabilizer
Heaters Gas Burners
Brass, rotatable air regulator for safe and easy adjustment of flame.
  • Premium quality Bunsen burner, suitable for a variety of laboratory applications
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Die-cast zinc alloy base
Lab quality gas burner with an nickel-plated burner mixing tube, brass flame stabilizer for safety, and a die cast zinc alloy base. Burner tube diameter is 13 mm. Overall size: 6.3"H × 3" base Ø.