Metal Mini Boss Head
Supplier: Wards
CH0662
470344-134EA 11.05 USD
470344-134
Metal Mini Boss Head
Clamps Bossheads
Metal mini boss head with a 15 mm diameter, providing secure attachment and flexibility for various lab apparatus.
  • Boss head with threaded screws capable of attaching to rods 15 mm in diameter
  • Holds lab equipment at a right angle
  • Strong, durable die casted alloy prevents flexure or warping
  • Perfect tool for a lab setup that is short on space
  • Great for classroom lab setups
Bosshead capable of holding lab equipment on a stand. Threaded screws can fit rods up to 13 mm, and holds equipment orthogonally. The die casted, all metal construction prevents damage to the boss head and apparatus. This boss head is perfect for small or tightly-spaced setups in a classroom or professional lab.