Set of 12 Drilled Balls

Supplier: Wards
PH0109CB
470344-120EA 105.45 USD
470344-120
Set of 12 Drilled Balls
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Multi-purpose drilled ball set ideal for pendulum experiments but can be used to meet a variety of lab needs.

  • Balls are 25 mm in diameter
  • Each ball has a 3 mm diameter hole

Contains two balls each of wood, aluminium, steel, cork, brass and lead.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR