Eisco Latex Dropper Bulbs
Supplier: Wards
CH0232DPK72 CH0232CPK72
470344-042EA 43.25 USD
470344-042 470344-007
Eisco Latex Dropper Bulbs
Pipettes Pipette Bulbs
Bulbs made from high-quality latex fit pipettes and medicine droppers measuring 6 to 8 mm in diameter.
  • Pack of 72 dropper bulbs made of high quality latex rubber
  • Excellent for science classrooms and professional laboratories
Latex dropper bulbs fit glass pipettes.