Eisco® Buzzer in Vacuum Jar Kit
Supplier: Wards
PH0178
470343-978EA 88.5 USD
470343-978
Eisco® Buzzer in Vacuum Jar Kit
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Airtight chamber reveals how sound disappears without air.
  • Built-in buzzer activates via external switch
  • Clear, impact-resistant acrylic construction
  • Compatible with standard hand vacuum systems
This apparatus helps students explore the fundamental role of air in sound transmission. Once sealed and evacuated, the chamber isolates the buzzer's vibrations from the surrounding environment, creating a silent visual paradox. Ideal for physics demonstrations, the setup encourages critical thinking and discussion. A vacuum pump is required for full functionality and must be sourced separately.