Eisco Amber Glass Dropping Bottle, 180 ml
Supplier: Wards
CH0187H
470343-872EA 5.7 USD
470343-872
Eisco Amber Glass Dropping Bottle, 180 ml
Bottles Dropping and Dispensing Bottles
Perfect for transferring and mixing stains, indicators, oils, or other aqueous solutions.
  • High quality dropping bottle made of soda glass
  • Amber color glass appropriate for light-sensitive contents
  • Includes a screw top cap with built in glass pipette dropper and rubber bulb
  • 180 ml (6 oz.) capacity
Suitable for a variety of applications in the classroom or laboratory. Bottle includes a screw top cap with built in glass pipette dropper and rubber bulb. Bottle (with screw cap) measures 6" tall and 2.2" in diameter.